Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Accs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290782.1 | NP_001277711.1 | 3 | 508 | ||
NM_183220.3 | NP_899043.1 | 3 | 563 | |||
GenBank mRNA | AK011867.1 | - | 3 | 412 | ||
AK040509.1 | - | 3 | 405 | |||
BC039569.1 | - | 3 | 540 | 1449080 |