Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177845.4 | NP_808513.2 | 13 | 1719 | ||
GenBank mRNA | AB195277.1 | - | 13 | 1622 | ||
AK049063.1 | - | 12 | 2252 | |||
BC060600.1 | - | 10 | 1665 | |||
BC066085.1 | - | 13 | 1720 | |||
BC137899.1 | - | 13 | 1496 | 8860918 | ||
BC145020.1 | - | 14 | 1607 |