Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002863.4 | NP_002854.3 | 2 | 412 | ||
GenBank mRNA | AF046785.1 | - | 2 | 285 | ||
AF066858.1 | - | 2 | 285 | |||
AK225904.1 | - | 2 | 365 | |||
AK309333.1 | - | 2 | 338 | |||
AK313206.1 | - | 2 | 326 | |||
AL134906.1 | - | 2 | 398 | |||
BC009895.2 | - | 2 | 355 | 2967205 | ||
BC065011.1 | - | 2 | 352 | |||
BC082229.1 | - | 2 | 356 | 4053354 | ||
BC095850.1 | - | 2 | 372 | 30343100 | ||
BC110791.1 | - | 2 | 354 | 6051020 | ||
M14636.1 | - | 2 | 398 | |||
Y15233.1 | - | 2 | 336 |