Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYGM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164716.1 | NP_001158188.1 | 16 | 2751 | ||
GenBank mRNA | AF066859.1 | - | 18 | 2198 | ||
AK056607.1 | - | 16 | 2010 | |||
AK057547.1 | - | 1 | 1550 | |||
AK299376.1 | - | 14 | 1538 | |||
AK299461.1 | - | 18 | 2217 | |||
AK314474.1 | - | 18 | 2202 | |||
BC126392.1 | - | 18 | 2597 | 8992108 | ||
BC130514.1 | - | 18 | 2597 | 40146545 | ||
X03031.1 | - | 2 | 175 |