Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shisa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033415.3 | NP_001028587.1 | 2 | 1133 | ||
GenBank mRNA | AK077061.1 | - | 2 | 374 | ||
AK143290.1 | - | 2 | 1134 | |||
BC120534.1 | - | 2 | 641 | 40129457 | ||
BC120536.1 | - | 2 | 641 | 40129459 |