Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pate2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033421.3 | NP_001028593.1 | 3, 4 | 261 | ||
GenBank mRNA | AK136651.1 | - | 3, 4 | 262 | ||
AK136739.1 | - | 1, 2 | 971 |