Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212487.4 | NP_997652.4 | 4, 5 | 807 | ||
GenBank mRNA | AK146611.1 | - | 4, 5 | 810 | ||
AY651827.1 | - | 4, 5 | 632 | |||
BC119600.1 | - | 4, 5 | 744 | |||
BC119601.1 | - | 4, 5 | 744 | |||
BC127241.1 | - | 4, 5 | 744 | |||
BC127618.1 | - | 4, 5 | 744 |