Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175380.5 | NP_780589.3 | 2 | 436 | ||
GenBank mRNA | AK050572.1 | - | 2 | 329 | ||
AK075845.1 | - | 2 | 278 | |||
AK145475.1 | - | 2 | 291 | |||
AK166662.1 | - | 2 | 288 | |||
AK172886.1 | - | 2 | 381 | |||
BC037729.1 | - | 2 | 299 | |||
BC138976.1 | - | 2 | 264 | 8861998 | ||
BC138980.1 | - | 2 | 264 | 8862002 |