Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030981.2 | NP_112243.1 | 6 | 602 | ||
XM_017018378.1 | XP_016873867.1 | 5 | 769 | |||
GenBank mRNA | AF092437.1 | - | 6 | 520 | ||
AJ245875.1 | - | 5 | 509 | |||
AK292086.1 | - | 6 | 570 | |||
AL136635.1 | - | 6 | 554 | |||
BC028331.1 | - | 6 | 553 | |||
BC071169.1 | - | 6 | 541 | 6026121 | ||
CR533462.1 | - | 5 | 507 |