Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181470.4 | NP_852135.1 | 4 | 473 | ||
GenBank mRNA | AK011491.1 | - | 4 | 459 | ||
BC067062.1 | - | 4 | 473 | 30538957 | ||
BC069962.1 | - | 4 | 467 | 5038459 | ||
U01139.1 | - | 1 | 48 |