Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181729.2 | NP_859418.1 | 3165 | |||
GenBank mRNA | AJ010752.1 | - | 7 | 789 | ||
BC120907.2 | - | 24 | 3053 | 40087870 |