Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291212.1 | NP_001278141.1 | 1 | 669 | ||
NM_001291213.1 | NP_001278142.1 | 1 | 669 | |||
NM_011586.3 | NP_035716.1 | 1 | 669 | |||
GenBank mRNA | AB026497.1 | - | 1 | 639 | ||
AK146852.1 | - | 2 | 749 | |||
BC138366.1 | - | 1 | 611 | 8861386 | ||
BC145257.1 | - | 1 | 611 |