Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291212.1 | NP_001278141.1 | 3 | 1152 | ||
NM_001291213.1 | NP_001278142.1 | 3 | 1260 | |||
NM_001291214.1 | NP_001278143.1 | 2 | 119 | |||
NM_001291215.1 | NP_001278144.1 | 3 | 155 | |||
NM_011586.3 | NP_035716.1 | 2 | 1116 | |||
GenBank mRNA | AB026497.1 | - | 2 | 1086 | ||
AK137574.1 | - | 2 | 107 | |||
AK147584.1 | - | 3 | 104 | |||
AK171342.1 | - | 2 | 116 | |||
BB851963.1 | - | 2 | 121 | |||
BC138366.1 | - | 3 | 1094 | 8861386 | ||
BC145257.1 | - | 3 | 1202 |