Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198119.2 | NP_932787.1 | 6 | 916 | ||
GenBank mRNA | AK048678.1 | - | 6 | 909 | ||
AK078176.1 | - | 6 | 917 | |||
BC116884.1 | - | 4 | 708 | 40126203 | ||
BC116886.1 | - | 4 | 708 | 40126205 |