Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raet1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198193.2 | NP_937836.1 | 5 | 677 | ||
GenBank mRNA | AK012289.1 | - | 5 | 677 | ||
AK131854.1 | - | 7 | 980 | |||
AY054973.1 | - | 3 | 543 | |||
AY056835.1 | - | 3 | 543 | |||
BC132308.1 | - | 3 | 580 | 40130585 | ||
FJ594067.1 | - | 6 | 781 |