Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163267.1 | NP_001156739.1 | 2 | 167 | ||
GenBank mRNA | BC057135.1 | - | 2 | 167 | ||
BC147530.1 | - | 2 | 156 | 9056521 | ||
BC147540.1 | - | 2 | 156 | 9056531 | ||
BC150978.1 | - | 2 | 157 | 9087891 | ||
BC150979.1 | - | 2 | 156 | 9087892 |