Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033449.1 | NP_001028621.1 | 6 | 441 | ||
NM_001037812.2 | NP_001032901.1 | 6 | 400 | |||
GenBank mRNA | AK133112.1 | - | 7 | 425 | ||
AK161697.1 | - | 6 | 443 | |||
AK167733.1 | - | 6 | 402 | |||
AK170118.1 | - | 6 | 381 | |||
BC147243.1 | - | 6 | 420 | 9056304 | ||
BC147244.1 | - | 6 | 420 | 9056305 | ||
BF722386.1 | - | 2 | 87 |