Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033449.1 | NP_001028621.1 | 8 | 688 | ||
NM_001037812.2 | NP_001032901.1 | 8 | 647 | |||
GenBank mRNA | AK133112.1 | - | 9 | 672 | ||
AK161697.1 | - | 8 | 691 | |||
AK167733.1 | - | 8 | 649 | |||
AK170118.1 | - | 8 | 628 | |||
BC147243.1 | - | 8 | 667 | 9056304 | ||
BC147244.1 | - | 8 | 667 | 9056305 | ||
BF722386.1 | - | 4 | 334 |