Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rc3h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024952.2 | NP_001020123.1 | 2 | 585 | ||
GenBank mRNA | AK139648.1 | - | 2 | 557 | ||
AK173335.1 | - | 2 | 531 | |||
AY948287.1 | - | 2 | 526 | |||
BC138663.1 | - | 1 | 309 | 8861684 |