Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mup21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009550.2 | NP_001009550.1 | 3 | 354 | ||
GenBank mRNA | BC141241.1 | - | 3 | 349 | 9055853 | |
EU882236.1 | - | 3 | 265 |