Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xkr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201368.1 | NP_958756.1 | 3 | 1183 | ||
GenBank mRNA | AK029920.1 | - | 3 | 1183 | ||
AY534254.1 | - | 3 | 1183 | |||
BC057035.1 | - | 3 | 1119 | 5698137 |