Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028234.1 | NP_082510.1 | 5 | 724 | ||
GenBank mRNA | AK014289.1 | - | 5 | 713 |