Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3glct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081204.1 | NP_001074673.1 | 6 | 458 | ||
GenBank mRNA | AK089440.1 | - | 6 | 458 | ||
AK145082.1 | - | 6 | 432 |