Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005055.4 | NP_005046.2 | 1 | 295 | ||
NM_032645.4 | NP_116034.2 | 1 | 295 | |||
XM_005253042.3 | XP_005253099.1 | 1 | 253 | |||
XM_005253043.3 | XP_005253100.1 | 1 | 253 | |||
XM_011520252.1 | XP_011518554.1 | 1 | 253 | |||
GenBank mRNA | AF449218.1 | - | 1 | 295 | ||
AK310002.1 | - | 1 | 253 | |||
BC004196.2 | - | 1 | 260 | 3530551 | ||
KU178300.1 | - | 1 | 81 | |||
KU178301.1 | - | 1 | 81 | |||
KU178302.1 | - | 1 | 81 | |||
KU178303.1 | - | 1 | 81 | |||
KU178304.1 | - | 1 | 81 | |||
Z33905.1 | - | 1 | 288 |