Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Colq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009937.2 | NP_034067.2 | 14 | 1176 | ||
GenBank mRNA | AK134212.1 | - | 14 | 1180 | ||
BC107386.2 | - | 14 | 1110 | 40057706 | ||
BC107387.1 | - | 13 | 973 | 40057715 |