Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smok3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126045.1 | NP_001119517.1 | 2 | 829 | ||
NM_001168602.1 | NP_001162073.1 | 2 | 982 | |||
GenBank mRNA | AJ245454.1 | - | 2 | 982 | ||
BC150960.1 | - | 829 | 9087871 |
Gene Symbol: Smok3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039889.3 | NP_001034978.1 | 2 | 1003 | ||
GenBank mRNA | AK133180.1 | - | 2 | 1004 |