Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lca5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001492.2 | NP_001001492.2 | 5 | 763 | ||
GenBank mRNA | AK029554.1 | - | 5 | 757 | ||
AK133316.1 | - | 5 | 763 | |||
BC141285.1 | - | 4 | 613 | 9055897 | ||
BC145611.1 | - | 4 | 613 | 9054177 |