Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt1a6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145079.3 | NP_659545.2 | 331 | |||
GenBank mRNA | AK162514.1 | - | 327 | |||
AY227197.1 | - | 1 | 299 | |||
BC012716.1 | - | 325 | ||||
BC069940.1 | - | 321 | 5249697 | |||
U09930.1 | - | 1 | 213 | |||
U16818.1 | - | 331 |
Gene Symbol: Ugt1a6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201410.3 | NP_958812.3 | 2 | 326 | ||
GenBank mRNA | AY227198.1 | - | 1 | 282 | ||
BC026561.1 | - | 2 | 306 | 5050488 |