Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgprb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207538.1 | NP_997421.1 | 1 | 707 | ||
GenBank mRNA | BC120849.1 | - | 1 | 740 | 40129772 | |
BC132003.1 | - | 1 | 741 | 40130280 |
Set Membership: |
G-protein coupled receptor |