Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: V1rd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207619.2 | NP_997502.2 | 1 | 641 | ||
GenBank mRNA | BC107173.1 | - | 1 | 641 | 40050592 | |
BC107174.1 | - | 1 | 641 | 40050594 |
Gene Symbol: V1rd18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207618.2 | NP_997501.2 | 1 | 661 | ||
GenBank mRNA | BC147785.1 | - | 1 | 661 | 9087504 | |
BC147787.1 | - | 1 | 661 | 9087506 |