Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPN1LW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020061.5 | NP_064445.2 | 4 | 766 | ||
GenBank mRNA | AW950066.1 | - | 1 | 130 | ||
BQ637240.1 | - | 3 | 440 | |||
CR749814.1 | - | 4 | 724 | |||
EL949562.1 | - | 2 | 275 |
Set Membership: |
G-protein coupled receptor |