Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2afy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207000.2 | NP_996883.1 | 5 | 635 | ||
GenBank mRNA | AF336305.1 | - | 5 | 635 | ||
AK018355.1 | - | 5 | 701 | |||
AK032636.1 | - | 5 | 718 | |||
AK078464.1 | - | 5 | 705 | |||
AK165020.1 | - | 5 | 682 | |||
BC045140.1 | - | 5 | 676 | 6307273 | ||
BC046794.1 | - | 5 | 704 | 5717897 | ||
BC107279.1 | - | 4 | 542 | 40052731 | ||
BC107280.1 | - | 4 | 542 | 40052733 | ||
BC127144.1 | - | 4 | 542 | 40052734 |