Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Specc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029936.3 | NP_001025107.1 | 2 | 765 | ||
NM_001281818.1 | NP_001268747.1 | 4 | 982 | |||
GenBank mRNA | AK046469.1 | - | 4 | 967 | ||
AK050982.1 | - | 2 | 766 | |||
AK143663.1 | - | 4 | 982 | |||
AK220465.1 | - | 2 | 692 |