Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033775.3 | NP_001028947.1 | 4, 5 | 864 | ||
GenBank mRNA | AK133415.1 | - | 4, 5 | 865 | ||
BC147608.1 | - | 4, 5 | 882 | 9056598 | ||
BC157912.1 | - | 4, 5 | 882 | 9088129 |