Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290751.1 | NP_001277680.1 | 6 | 749 | ||
NM_025868.4 | NP_080144.1 | 8 | 1049 | |||
GenBank mRNA | AK009759.1 | - | 7 | 737 | ||
AK075750.1 | - | 8 | 1051 | |||
AK170587.1 | - | 7 | 746 | |||
BC013544.1 | - | 7 | 731 | 4242418 | ||
BC110994.1 | - | 7 | 732 | 5349078 |