Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204747.1 | NP_001191676.1 | 10 | 1274 | ||
NM_002913.4 | NP_002904.3 | 10 | 1274 | |||
XM_011513730.1 | XP_011512032.1 | 9 | 1196 | |||
XM_011513731.1 | XP_011512033.1 | 10 | 1274 | |||
XM_017008528.1 | XP_016864017.1 | 9 | 1196 | |||
GenBank mRNA | AF040250.1 | - | 10 | 1179 | ||
AK291612.1 | - | 10 | 1255 | |||
BC035297.1 | - | 10 | 1225 | 3923331 | ||
BC051751.1 | - | 10 | 1244 | 6070144 | ||
BC051786.2 | - | 10 | 1255 | 5521245 | ||
L23320.1 | - | 10 | 1340 | |||
X69821.1 | - | 6 | 803 |