Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033782.3 | NP_001028954.1 | 2 | 247 | ||
GenBank mRNA | AK133295.1 | - | 2 | 248 | ||
BC147594.1 | - | 2 | 132 | 9056584 |