Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206691.1 | NP_001193620.1 | 9 | 1079 | ||
NM_032491.5 | NP_115880.2 | 5 | 745 | |||
NM_213594.2 | NP_998759.1 | 9 | 1326 | |||
GenBank mRNA | AB044245.1 | - | 9 | 1044 | ||
AB095365.1 | - | 9 | 1074 | |||
AB095366.1 | - | 9 | 1326 | |||
AF332192.1 | - | 5 | 739 | |||
AK074913.1 | - | 6 | 657 | |||
AK291445.1 | - | 9 | 1311 | |||
AK302837.1 | - | 9 | 1019 | |||
AK315698.1 | - | 5 | 725 | |||
AL833921.1 | - | 2 | 149 | |||
AY102009.1 | - | 9 | 979 | |||
BC028582.2 | - | 5 | 748 | 4836964 | ||
BC030644.1 | - | 5 | 720 | 4825079 |