Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6ap1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145879.1 | NP_001139351.1 | 5, 6 | 531 | ||
GenBank mRNA | CX238982.1 | - | 5, 6 | 472 |