Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RHD SIRNA Assays ›
Gene Symbol:
RHD
Gene Name:
Rh blood group D antigen
Gene Aliases:
CD240D, DIIIc, RH, RH30, RHCED, RHDVA(TT), RHDel, RHPII, RHXIII, Rh4, RhDCw, RhII, RhK562-II, RhPI
Chromosome Location:
Chr.1: 25272393 - 25330445 on Build GRCh38
UniGene ID:
Hs.449968
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s12017
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RHD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127691.2
NP_001121163.1
4
655
NM_001282867.1
NP_001269796.1
4
651
NM_001282868.1
NP_001269797.1
4
655
NM_001282869.1
NP_001269798.1
4
655
NM_001282870.1
NP_001269799.1
4
655
NM_001282871.1
NP_001269800.1
4
655
NM_001282872.1
NP_001269801.1
4
655
NM_016124.4
NP_057208.2
4
655
XM_017002015.1
XP_016857504.1
4
748
GenBank mRNA
AB018966.1
-
4
593
AB018967.1
-
4
593
AB018968.1
-
4
593
AB018969.1
-
4
593
AB037270.1
-
4
593
AB046420.1
-
4
593
AB049754.1
-
4
593
AF037626.1
-
4
589
AF177938.1
-
4
593
AF510069.1
-
4
636
AF510070.1
-
4
636
AJ704215.2
-
4
593
AK297151.1
-
4
748
AM183924.3
-
4
593
AY449381.1
-
4
593
AY449382.1
-
4
593
AY449383.1
-
4
593
AY449384.1
-
4
593
AY449385.1
-
4
593
AY751491.1
-
4
593
AY751492.1
-
4
593
AY751493.1
-
4
593
AY751494.1
-
4
593
AY751495.1
-
4
593
AY751496.1
-
4
593
BC139922.1
-
4
643
40148684
DQ451878.1
-
4
612
EU557240.1
-
4
596
EU557241.1
-
4
596
EU661905.1
-
4
533
EU661906.1
-
4
533
EU661907.1
-
1
86
FN555129.1
-
1
107
FR745439.1
-
4
593
FR745440.1
-
4
593
FR745441.1
-
4
593
HE657775.1
-
4
593
JN635688.1
-
4
593
JQ065733.1
-
4
593
JQ424879.1
-
4
651
KT240043.1
-
4
593
L08429.1
-
4
593
LC053447.1
-
4
593
LN714468.1
-
4
593
LN824132.1
-
4
593
S57971.1
-
4
593
S70174.1
-
4
593
S73913.1
-
4
593
S78509.1
-
4
593
S82449.1
-
4
593
X63094.1
-
4
651
X63097.1
-
4
609
Back To Top
More Information
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
organic cation transport
nitrogen utilization
ammonium transmembrane transport
ammonium transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0