Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znhit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005223.2 | NP_001005223.1 | 5 | 429 | ||
GenBank mRNA | AK002888.1 | - | 5 | 421 | ||
AK003721.1 | - | 5 | 428 | |||
AK004259.1 | - | 5 | 431 | |||
AK014350.1 | - | 5 | 440 | |||
BC089554.1 | - | 5 | 517 | 30300471 |