Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002929.2 | NP_002920.1 | 7 | 1543 | ||
XM_017020684.1 | XP_016876173.1 | 7 | 3210 | |||
GenBank mRNA | AK289912.1 | - | 7 | 1667 | ||
BC140945.1 | - | 7 | 1508 | 9056757 | ||
BM702747.1 | - | 2 | 132 | |||
BX647503.1 | - | 1 | 631 | |||
L77503.1 | - | 7 | 1450 | |||
U63973.1 | - | 7 | 1543 |