Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256820.1 | NP_001243749.1 | 3 | 244 | ||
NM_001256821.1 | NP_001243750.1 | 4 | 271 | |||
NM_006912.5 | NP_008843.1 | 4 | 393 | |||
GenBank mRNA | AF084462.1 | - | 4 | 304 | ||
AF493923.1 | - | 3 | 190 | |||
AK298768.1 | - | 3 | 244 | |||
AK314239.1 | - | 4 | 369 | |||
BC104186.2 | - | 4 | 264 | 40030939 | ||
BC104187.2 | - | 4 | 264 | 40030940 | ||
BI916790.1 | - | 4 | 254 | |||
BM546985.1 | - | 4 | 387 | |||
BQ918446.1 | - | 4 | 271 | |||
CR407639.1 | - | 3 | 190 | |||
U71203.1 | - | 4 | 325 | |||
U78165.1 | - | 4 | 325 | |||
Y07566.1 | - | 4 | 335 |