Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177652.2 | NP_001171123.1 | 3 | 432 | ||
GenBank mRNA | AB256662.1 | - | 3 | 419 | ||
BC019499.1 | - | 2 | 1211 | |||
BC038040.1 | - | 2 | 1230 | |||
BC055452.1 | - | 3 | 434 | |||
BC055924.1 | - | 3 | 420 | |||
BC069936.1 | - | 3 | 420 | |||
BC087541.1 | - | 3 | 427 | |||
BC108376.1 | - | 3 | 426 | |||
BF301854.1 | - | 3 | 442 | |||
BG865695.1 | - | 3 | 431 |