Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5891 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034904.2 | NP_001030076.1 | 4 | 775 | ||
GenBank mRNA | AK139650.1 | - | 4 | 775 | ||
AK163219.1 | - | 646 |
Gene Symbol: Gm10662 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201364.1 | NP_001188293.1 | 4 | 773 |