Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081415.1 | NP_001074884.1 | 2 | 1169 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | W83690.1 | - | 88 | Chr. - - [-] on Build |