Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RNH1 SIRNA Assays ›
Gene Symbol:
RNH1
Gene Name:
ribonuclease/angiogenin inhibitor 1
Gene Aliases:
RAI, RNH
Chromosome Location:
Chr.11: 494512 - 507283 on Build GRCh38
UniGene ID:
Hs.530687
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s12084
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RNH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002939.3
NP_002930.2
4
608
NM_203383.1
NP_976317.1
4
540
NM_203384.1
NP_976318.1
3
367
NM_203385.1
NP_976319.1
3
435
NM_203386.2
NP_976320.1
4
498
NM_203387.2
NP_976321.1
4
566
NM_203388.2
NP_976322.1
3
325
NM_203389.2
NP_976323.1
3
393
XM_011520255.1
XP_011518557.1
4
543
XM_011520257.2
XP_011518559.1
4
791
XM_011520258.2
XP_011518560.1
4
759
XM_011520259.2
XP_011518561.1
3
308
XM_011520260.2
XP_011518562.1
4
467
XM_011520261.2
XP_011518563.1
4
541
XM_011520262.2
XP_011518564.1
4
724
XM_011520263.1
XP_011518565.1
3
275
XM_017018106.1
XP_016873595.1
4
608
GenBank mRNA
AB209196.1
-
3
1215
AK308340.1
-
4
495
AK310173.1
-
4
478
AK311379.1
-
3
355
AL161967.1
-
2
1543
AL534410.3
-
3
382
AL539148.3
-
3
410
BC000677.2
-
3
295
3349381
BC003506.1
-
3
308
2959514
BC011186.1
-
2
93
BC011500.1
-
3
246
4155804
BC014629.2
-
3
295
3029730
BC024037.1
-
3
308
2959514
BC047730.1
-
4
587
5788220
BG716555.1
-
3
383
BG720935.1
-
3
308
BG752774.1
-
4
506
BM019570.1
-
3
368
BM465975.1
-
4
444
BM562998.1
-
3
366
BQ927358.1
-
3
254
BQ954409.1
-
3
361
DC296330.1
-
4
478
M22414.1
-
3
256
M36717.1
-
4
495
X13973.1
-
4
495
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA catabolic process
negative regulation of catalytic activity
regulation of angiogenesis
protein binding
ribonuclease inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0