Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlrad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081272.1 | NP_001074741.1 | 4 | 744 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG964589.1 | - | 666 | Chr. - - [-] on Build |