Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002944.2 | NP_002935.2 | 38 | 6307 | ||
XM_006715548.3 | XP_006715611.1 | 39 | 6399 | |||
XM_011536049.2 | XP_011534351.1 | 39 | 6445 | |||
XM_011536050.2 | XP_011534352.1 | 39 | 6442 | |||
XM_011536051.2 | XP_011534353.1 | 38 | 6418 | |||
XM_011536052.2 | XP_011534354.1 | 39 | 6403 | |||
XM_011536053.2 | XP_011534355.1 | 39 | 6271 | |||
XM_011536054.2 | XP_011534356.1 | 39 | 6446 | |||
XM_017011172.1 | XP_016866661.1 | 38 | 6376 | |||
XM_017011173.1 | XP_016866662.1 | 38 | 6475 | |||
GenBank mRNA | BC144556.1 | - | 36 | 5489 | ||
M13880.1 | - | 5 | 551 | |||
M34353.1 | - | 38 | 6314 |