Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glrx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023140.4 | NP_075629.2 | 4 | 411 | ||
GenBank mRNA | AF118650.1 | - | 4 | 409 | ||
AK010354.1 | - | 4 | 432 | |||
AK017371.1 | - | 4 | 412 | |||
AK051468.1 | - | 4 | 405 | |||
AK147158.1 | - | 4 | 392 | |||
AK152446.1 | - | 4 | 400 | |||
AK152634.1 | - | 4 | 400 | |||
AK155190.1 | - | 4 | 398 | |||
AK167672.1 | - | 4 | 446 | |||
AK186287.1 | - | 1 | 26 | |||
AK215855.1 | - | 1 | 35 | |||
BC033506.1 | - | 4 | 396 | 2651132 | ||
BC087885.1 | - | 4 | 404 | 30303379 |